This item is now discontinued. Please browse similar items
nVent Hoffman A10086CHFL Type 4 Quarter-Turn Fast Latch Steel Junction Box
Representative image only.
nVent Hoffman Mfr # A10086CHFL – Item # WZ-10502-58
This enclosure provides the convenience of quarter-turn latching with an aesthetic, easy-to-maintain light gray textured finish
  • Quarter-turn latch with screwdriver slot for easy access
  • Oil-resistant, seamless foam-in-place gasket
  • Available in 14 or 16 gauge steel
  • Seams continuously welded and ground smooth
  • Continuous hinge
  • Bonding provision on door
  • Weld nuts provided for mounting optional panels and terminal block kits in size 6” x 4” (152 x 102 mm) and larger
  • Ratings: NEMA Type 4, 12; IP66
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSteel
  • Inside Width (in)8
  • Inside Height (in)10
  • Inside Depth (in)6
  • DescriptionType 4 Quarter-Turn Fast Latch Steel Junction Box
  • Warranty1 Year
1 Year WarrantyCanadian Standards AssociationIP 66 Rated
Key Features
  • Quarter-turn latch with screwdriver slot for easy access
  • Oil-resistant, seamless foam-in-place gasket
  • Available in 14 or 16 gauge steel
  • Seams continuously welded and ground smooth
  • Continuous hinge
  • Bonding provision on door
  • Weld nuts provided for mounting optional panels and terminal block kits in size 6” x 4” (152 x 102 mm) and larger
  • Ratings: NEMA Type 4, 12; IP66