This item is now discontinued. Please browse similar items
PCRmax qPCR Plate 96-Well white for RLC 480, low profile, half skirted & self-adhesive sealing film, 50/cs
Representative image only.
PCRmax Item # WZ-67104-47
Improve your heat transfer and cycle times
  • Constructed of high-quality polypropylene for optimal clarity and solvent resistance
  • Certified free of RNase, DNase, endotoxins, and human DNA
  • Suitable for use in most thermal cyclers
  • Choose from 96- and 384-well plates
  • Compatible with Roche® LightCycler®
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (mL)0.15
  • Plate TypeLow profile, half skirt with sealing foil
  • Plate ColorWhite
  • MaterialPP
  • DescriptionqPCR Plate 96-Well white for RLC 480, low profile, half skirted & self-adhesive sealing film, 50/cs
Key Features
  • Constructed of high-quality polypropylene for optimal clarity and solvent resistance
  • Certified free of RNase, DNase, endotoxins, and human DNA
  • Suitable for use in most thermal cyclers
  • Choose from 96- and 384-well plates
  • Compatible with Roche® LightCycler®
More About this Item
The qPCR plates feature ultrathin walls for accelerated heat transfer and cycle times. The 96-well plates are available with either 0.15- or 0.2-mL well volume; skirted, half-skirted, or unskirted; and low or standard profile well. The 384-well plate features a 40 µl well volume, 25 µl sample volume, and is skirted. The plates are designed to fit most major thermal cyclers and are autoclavable.