This item is now discontinued. Please browse similar items
PCRmax qPCR Plate 384-Well white for RLC 480, full skirted, 50/cs
Representative image only.
PCRmax Item # WZ-67104-77
Improve your heat transfer and cycle times
  • Constructed of high-quality polypropylene for optimal clarity and solvent resistance
  • Certified free of RNase, DNase, endotoxins, and human DNA
  • Suitable for use in most thermal cyclers
  • Choose from 96- and 384-well plates
SPECS & DESCRIPTION
Specifications & Description
Key Features
  • Constructed of high-quality polypropylene for optimal clarity and solvent resistance
  • Certified free of RNase, DNase, endotoxins, and human DNA
  • Suitable for use in most thermal cyclers
  • Choose from 96- and 384-well plates
More About this Item
The qPCR plates feature ultrathin walls for accelerated heat transfer and cycle times. The 96-well plates are available with either 0.15- or 0.2-mL well volume; skirted, half-skirted, or unskirted; and low or standard profile well. The 384-well plate features a 40 µl well volume, 25 µl sample volume, and is skirted. The plates are designed to fit most major thermal cyclers and are autoclavable.