This item is now discontinued. Please browse similar items
PCRmax DNA Specific Detection Kit for C. capreolus, C. elaphus, and D. dama (venison), without mastermix
Representative image only.
PCRmax Item # WZ-93945-64
Quickly and accurately verify the speciation in the food source
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Great value at 100 tests per kit
SPECS & DESCRIPTION
Specifications & Description
  • Test TypeDNA
  • Number of Tests Included100
  • Mastermix IncludedNo
  • DescriptionSpecific Detection Kit for C. capreolus, C. elaphus, and D. dama (venison), without mastermix
Includes
Lyophilized primer and probe mix, positive control for standard curve, internal extraction control DNA, endogenous control, and RNAse/DNAse-free water.
Key Features
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Great value at 100 tests per kit
More About this Item
Use the speciation kits to clarify sources and types of products where accurate results are critical. Order a complete kit with mastermix or select a kit without mastermix and use your own.