This item is now discontinued. Please browse similar items
PCRmax DNA Genesig CaMV 35S promoter and NOS terminator in GM Maize, with mastermix
Representative image only.
PCRmax Item # WZ-93940-04
Screen for genetically modified organisms (GMOs) in food, feed, and seeds
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
SPECS & DESCRIPTION
Specifications & Description
  • Test TypeDNA
  • Number of Tests Included150
  • Mastermix IncludedYes
  • DescriptionGenesig CaMV 35S promoter and NOS terminator in GM Maize, with mastermix
Includes
Lyophilized mastermix, lyophilized ROX, resuspension buffer, lyophilized primer and probe mix, positive control for standard curve, internal extraction control DNA, endogenous control, and RNAse/DNAse-free water.
Key Features
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
More About this Item
Kits with mastermix contain all the necessary components for fast and accurate results. To use your own mastermix, order kits without mastermix.