Parker Hannifin XV501P-4 1/4" NPT brass female/male manual ball valves
Representative image only.
Parker Hannifin Mfr # XV501P-4 – Item # WZ-30527-20
Don’t worry about leaks
  • Full flow design ensures maximum operating efficiency
  • Blow-out proof stems
PRODUCT OPTIONS
£41.58GBP / Each
£49.90GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/4" NPT(M)
  • Connection TypeNPT(F) x NPT(M)
  • Inlet Port1/4" NPT(F)
  • Cv6.3
  • Max Pressure (PSI)600
  • Max Temperature (° F)400
  • Max Temperature (° C)204
  • Wetted MaterialsBrass, PTFE
  • Working Pressure (PSI)150
  • Length (in)2.625
  • Description1/4" NPT brass female/male manual ball valves
Key Features
  • Full flow design ensures maximum operating efficiency
  • Blow-out proof stems
More About this Item
Speically-designed handle increases turning leverage and provides easy opening and closing. Brass valves are an ideal choice for use in water and air service lines. Stainless steel valves are ideal for use in corrosive environments such as chemical plants, refineries, and steel mills.