Parker Hannifin Flare compression pipe adapter, 1/4" OD x 3/8" NPT(M), PFA, Each
Representative image only.
Parker Hannifin Mfr # FAMS-46 – Item # WZ-30490-22
Leak-tight and high-pressure connection for corrosive chemical applications
PRODUCT OPTIONS
£50.61GBP / Each
£60.73GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPFA
  • Connection TypeFlare x NPT(M)
  • Thread size NPT(M) (port 2) (in)3/8
  • Compression Size / Tube Od (Port 1) (in)1/4
  • DescriptionFlare compression pipe adapter, 1/4" OD x 3/8" NPT(M), PFA, Each
More About this Item
PFA wetted parts and PVDF nuts provide high purity and excellent chemical resistance. Leaf-free design withstands pressures to 225 psi. Ideal for high-vibration or side-load stressed applications. Low dead volume reduces particle entrapment, limits bacterial growth in deionized water, and simplifies rinsing.