Parker Hannifin Compression Fitting Ferrule, 316 Stainless Steel, 1/2" OD; 1/EA
Representative image only.
Parker Hannifin Mfr # 8 TZ-SS – Item # WZ-03302-41
Get a secure seal at higher pressures
  • Constructed of durable stainless steel
PRODUCT OPTIONS
£9.82GBP / Each
£11.78GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialStainless Steel
  • Sterilization MethodEthylene oxide (EtO), autoclave, or gamma irradiation
  • Connection TypeCompression Fitting Ferrule
  • Compression Size / Tube Od (Port 1) (in)1/2
  • DescriptionCompression Fitting Ferrule, 316 Stainless Steel, 1/2" OD; 1/EA
Key Features
  • Constructed of durable stainless steel
More About this Item
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Ferrules are required to create a leak-tight seal.