Parker Hannifin Ball Valve, Polypropylene, 1/4'' OD Push-to-Connect x 1/4'' NPT(M)
Representative image only.
Parker Hannifin Mfr # PP4VMC4-MG – Item # WZ-01379-69
Create a secure and easy push-to-connect connection
  • Glass reinforced polypropylene (PP) body with a polysulfone ball and EPDM seals
  • NSF 51 listed
  • FDA compliant for contact with food
PRODUCT OPTIONS
£12.86GBP / Each
£15.43GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/4" Push to Connect
  • Connection TypeNPT(M) x Push to Connect
  • Inlet Port1/4" NPT(M)
  • Orifice (in)0.1875
  • Orifice (mm)4.76
  • Wetted MaterialsPolypropylene
  • DescriptionBall Valve, Polypropylene, Male Connector 1/4'' push-to-connect tube, 1/4'' NPT(M)
Key Features
  • Glass reinforced polypropylene (PP) body with a polysulfone ball and EPDM seals
  • NSF 51 listed
  • FDA compliant for contact with food
More About this Item
These Parker Hannifin polypropylene (PP) ball valves feature an all plastic design is leak free and corrosion resistant.
Valve, Ball, Polypropylene, Male Elbow 1/4'' push to connect tube, 1/4'' thread, 1/each