This item is now discontinued. Please browse similar items
Oscium WIPRY-PRO COMBO Combo Spectrum Analyzer / Power Meter
Representative image only.
Oscium Mfr # WIPRY-PRO COMBO – Item # WZ-20048-31
Ultra-portable RF tool measures pulse width, duty cycle and transmit on and off times
  • Enables your iPhone, iPad, or iPod touch to make power and frequency measurements
  • Frequency range allows you to visualize Wi-Fi, SSID’s, RSSI, Zigbee®, Bluetooth®, and remote sensor networks
  • Quickly and easily measure pulse width, duty cycle, and view transmit on and off times with precision power meter
  • Allows conducted measurements and non-RF related triggering
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Width (in)2.25
  • Depth (in)0.25
  • Height (in)1.5
  • DescriptionCombo Spectrum Analyzer / Power Meter
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • Enables your iPhone, iPad, or iPod touch to make power and frequency measurements
  • Frequency range allows you to visualize Wi-Fi, SSID’s, RSSI, Zigbee®, Bluetooth®, and remote sensor networks
  • Quickly and easily measure pulse width, duty cycle, and view transmit on and off times with precision power meter
  • Allows conducted measurements and non-RF related triggering
  • 1 year warranty
More About this Item
The WiPry-Pro Combo is a combination spectrum analyzer/power meter module that brings LCD RF power measurements into a graphical interface and displays the waveform of the amplitude with respect to time. This module allows you to capture, trigger, and record the actual power output similar to voltage on an oscilloscope. The touch-screen interface of the iPhone, iPad, and iPod touch makes this module extremely easy to use with any of these devices allowing you to leverage intuitive gestures like pinch to zoom. Lightweight, compact module fits is ideal for engineers and technicians conducting field measurements.
Setup and operation is simple—connect the module to your device (iPhone, iPad, or iPod touch), download the required free WiPry App from the Apple App Store (search for WiPry) that includes software, and begin taking measurements. The powerful software allows you to easily view, graph, and trend data in real time and download for future analysis. Module is compatible with iPhone 5S, iPhone 5C, iPhone 5, iPad Mini, iPad Air, iPad 4, iPod touch 5th generation.
Note: The Bluetooth word mark and logos are registered trademarks owned by Bluetooth SIG, Inc.