This item is now discontinued. Please browse similar items
Oscium OSC3055 Logic Harness, 5 Pin with SMD Grabbers
Representative image only.
Oscium Mfr # OSC3055 – Item # WZ-20044-23
Choose from our comprehensive line of Oscium iMSO-104 Oscilloscope Accessories and other Accessories for Oscilloscopes products.
  • Replacement digital logic harness for the iMSO-104 mixed signal oscilloscope
  • Connects the 4 digital channels and ground to your device under test
  • Connectors on the end of the wires accept SMD grabbers or standard 0.100 in. header pins
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionLogic Harness, 5 Pin with SMD Grabbers
Key Features
  • Replacement digital logic harness for the iMSO-104 mixed signal oscilloscope
  • Connects the 4 digital channels and ground to your device under test
  • Connectors on the end of the wires accept SMD grabbers or standard 0.100 in. header pins