Eagle Stainless Stainless Steel Dolly for 65L Storage Tank w/ Clip-Down Cover

Representative image only.
Eagle Stainless Mfr # KM-43 CHROME CASTERS – Item # WZ-07505-66
Safely transports storage tanks
  • Use with 07505-65
  • 3 month warranty

PRODUCT OPTIONS

£633.04GBP / Each
£759.65GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (Liters)65
  • MaterialStainless steel
  • Height (in)5.4
  • Diameter (in)17.2
  • Height (cm)13.7
  • Diameter (cm)43.7
  • DescriptionStainless SteelDolly for 65L Storage Tank w/ Clip-Down Cover
  • Warranty3 month
3 Month Warranty

Key Features

  • Use with 07505-65
  • 3 month warranty

More About this Item

The dollies are constructded of 304 stainless steel with chrome-plated casters. They feature four nylon swivel casters, two with brakes.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat