This item is now discontinued. Please browse similar items
Omni International 19-623 0.15 mm Garnet bead kit for Bead Ruptor 12
Representative image only.
Omni International Mfr # 19-623 – Item # WZ-04728-14
Prevent cross-contamination with disposable tubes
  • Ideal for use with Omni Bead Ruptor 12 homogenizer
SPECS & DESCRIPTION
Specifications & Description
  • Description0.15 mm Garnet bead kit for Bead Ruptor 12
Key Features
  • Ideal for use with Omni Bead Ruptor 12 homogenizer
More About this Item
Five different bead types allow processing of different samples in your Omni Bead Ruptor 12 homogenizer, 04728-01 (sold separately). Ceramic beads are ideal for soft animal, plant or human tissues. Metal beads are ideal for hard tissue types. Glass beads are ideal for processing microorganisms. Garnet beads are ideal for soil. Carbide beads are ideal for microbial and soil RNA processing.