This item is now discontinued. Please browse similar items
Omegadyne LCGD-25 Load Cell 25LB Capacity Mini Compression Load Cell
Representative image only.
Omegadyne Mfr # LCGD-25 – Item # WZ-93955-68
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lbs)25
  • Capacity (kg)11
  • Output2 mV/V nominal
  • DescriptionLoad Cell 25LB Capacity Mini Compression Load Cell
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Miniature industrial load cell These small low-profile, compression load cells have excellent long-term stability. An all stainless steel construction assures reliability in severe industrial environments. These cells are designed to operate by mounting on a flat surface, and they have a load button machined as an integral part of their basic design.