This item is now discontinued. Please browse similar items
Omegadyne LCFD-10 Load Cell 10LB Capacity Sub-Miniature Load Cell
Representative image only.
Omegadyne Mfr # LCFD-10 – Item # WZ-93955-55
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lbs)10
  • Capacity (kg)4.5
  • Accuracy=100 lb: ±0.15% FSO =250 lb: ±0.20% FSO
  • Output1 kg: 1.5 mV/V (nom) 5 to 1000 lb: 2 mV/V (nom)
  • DescriptionLoad Cell 10LB Capacity Sub-Miniature Load Cell
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Sub-miniature load cell
Small in size and capable of highly accurate readings, the LCFD Series comprises subminiature load cells for precision industrial applications. They are all stainless steel, measure both tension and compression
loads, and have male thread studs for load attachment. The exclusive internal design provides superior long-term stability and minimizes the effects of small off-axis loads.