This item is now discontinued. Please browse similar items
Omegadyne LC101-25 Load Cell 25LB Capacity S-Beam Ss Tension And Compression Load Cell
Representative image only.
Omegadyne Mfr # LC101-25 – Item # WZ-93955-33
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lbs)25
  • Capacity (kg)11
  • Output3 mV/V ±0.0075 mV/V
  • DescriptionLoad Cell 25LB Capacity S-Beam Ss Tension And Compression Load Cell
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
All Stainless Stell ''S'' Beams
  • FM Intrinsically Safe
  • All Stainless Steel for Harsh Industrial Applications
  • High Accuracy
  • 0.25% Interchangeability for Multiple Load Cell Applications
  • 5-Point Calibration Provided