Ohaus T32ME Defender Abs Plastic Dry Use Led Indicator

Representative image only.
Ohaus Mfr # T32ME – Item # WZ-90453-15
For dry Use applications
  • Steel housung with ABS faceplate
  • RS232 interface

PRODUCT OPTIONS

£155.00GBP / Each
£186.00GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Weighing Unitslb, oz, lb:oz, kg, g
  • Calibration TypeExternal
  • LanguagesEnglish
  • OutputRS-232
  • RS-232 InterfaceYes
  • Stabilization Time (Sec)2
  • Auto ShutoffYes
  • Height (in)5 1/5
  • Width (in)7 51/64
  • Depth (in)1 51/64
  • Display Height (in)51/64
  • DescriptionDefender Abs Plastic Dry Use Led Indicator

Key Features

  • Steel housung with ABS faceplate
  • RS232 interface

More About this Item

The T32ME indicator with its bright LED display is designed for floor scale and other installations where the display can be read from a distance and internal battery power are not required. The indicator features a painted steel housing with high-impact ABS faceplate, bi-directional RS232 interface. Painted steel mounting bracket included.

Backlite LCD display

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat