This item is now discontinued. View the direct replacement: WZ-91428-03 or browse similar items
Oakton Temp-100 Dual Input Thermocouple Thermometer with Boot
Representative image only.
Oakton Item # WZ-91427-40
Multi-data display shows both temperatures and the temperature differential simultaneously
  • 3-way hands-free operation.
  • Nonvolatile memory stores up to 1000 sets of readings
  • Rugged armor—built to withstand the toughest conditions.
  • Easy-to-use automatic field calibration ensures accurate readings.
  • Probes are sold separately
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermocouple
  • Number Of Probe Channels2
  • Types Of Probes AcceptedType E, Type J, Type K, Type T
  • Min Temperature (° F)-418
  • Max Temperature (° F)3272
  • Min Temperature (° C)-250
  • Max Temperature (° C)1800
  • Accuracy±0.1% of reading +0.7°F; ±0.25% of reading +2.0°F above -148°F [±0.1% of reading +0.4°C; ±0.25% of reading +1.0°C above -99.9°C]
  • Resolution0.1°; 1° above 999.9° and below -99.9°
  • Low Battery IndicatorYes
  • Display Type1.5\" Display, 4-digit LCD
  • Height (in)7
  • Depth (in)4
  • Battery Life400 hours
  • BatteryAA Battery (3)
  • DescriptionTemp-100 Dual Input Thermocouple Thermometer with Boot
  • Warranty3 year
3 Year Warranty
Key Features
  • 3-way hands-free operation.
  • Nonvolatile memory stores up to 1000 sets of readings
  • Rugged armor—built to withstand the toughest conditions.
  • Easy-to-use automatic field calibration ensures accurate readings.
  • Probes are sold separately
  • 3 year warranty
More About this Item
Choose the Oakton® Temp-100 thermometer for single temperature readings or simultaneous dual temperature readings. Meter accepts all type J, T, E, and K thermocouple probes with miniconnector. This robust and reliable meters work in any environment from the lab to the production floor. The sturdy armored construction protects against knocks and jars. Ergonomically designed for both large and small hands alike. The sealed keypad and ABS case meet IP54 standards for splash resistance. The meter has a large illuminated display, and a seven-button control panel that allows the user through easy-to-use menus to choose desired functionality.
Features include Min/Max, Hold, and average functions; °F/°C selection; offset; out-of-range alarm with audible/visible indicator; auto shutoff; and low-battery indicator. Store up to 1000 sets of readings in the nonvolatile memory. Take differential temperature measurements with the T1/T2 function.
The multidata LCD simultaneously indicates the readings of each probe, as well as the differential reading. For improved readability with rapidly changing temperatures, the meter resolution is
keypad selectable. Above 999.9°, the meter automatically switches from 0.1° resolution to 1° resolution, below –99°C the meter autoranges to 1°C resolution.
The Temp-100 is individually factory calibrated. Automatic field calibration is easily performed for oither a single- or dual-point calibration for each probe. Simply place the probe in a container packed with ice, fill with water, activate the calibration function, and the meter automatically recognizes the freezing point.
OAKTON’s exclusive “3-WAY HANDS-FREE” capability allows “hands-free” operation—use the built-in stand on a table top, attach the meter to metal objects through magnets inserted into
the armor boot, or hang from a pipe or belt using the hook-and-loop strap. Meter comes standard with the integral stand; the “hands-free” option adds magnets and strap (order separately)