Oakton pH Probe, Refillable, Flushable With Bulb Guard, PTFE DJ, Epoxy, BNC
Representative image only.
Oakton Item # WZ-35805-09
Outstanding value at an economical cost!
  • Compatible with all Oakton brand pH meters
  • Ideal for dirty samples
  • Flushable junction
  • Impact resistant
  • Double junction type
  • 1 year warranty
PRODUCT OPTIONS
£294.53GBP / Each
£353.44GBP (incl VAT)
Often Bought with…
Oakton Three-Speed, Battery-Operated Hygrothermograph
£813.02GBP / Each
£975.62GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsDirty Samples-flushable
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)0
  • Max pH (pH)12
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)110
  • Cable Length (m)1
  • DescriptionpH Probe, Refillable/Flushable PTFE DJ/Epoxy; BNC
  • Warranty1 year
1 Year Warranty
Key Features
  • Compatible with all Oakton brand pH meters
  • Ideal for dirty samples
  • Flushable junction
  • Impact resistant
  • Double junction type
  • 1 year warranty
More About this Item
Oakton combination electrodes are available in a variety of styles to meet the needs of your application. Epoxy electrodes are impact resistant and provide excellent durability for field applications. Glass electrodes withstand aggressive chemicals. Choose junction and tip style based on the needs of your application. All electrodes feature Ag/AgCl reference cells.