This item is now discontinued. We recommend the following as suggested replacements: WZ-35660-70

Oakton pH 620 Waterproof Handheld Meter Only

Representative image only.
Oakton Item # WZ-35418-22
Expanded range, resolution and accuracy, increased memory, and advanced communications!
  • Measure pH or ISE with 0.001 maximum resolution
  • Out-of-date or unperformed calibrations are now things of the past!
  • Don't waste time looking for those cables!
  • Use in dirty and wet environments
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 6 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min ISE0 to 19999 ppm
  • ISE Resolution0.001 ppm; 0.001 molar
  • ISE Accuracy±0.5% of reading
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Min Temperature (° F)14
  • Max Temperature (° F)230
  • Temperature compensationAutomatic or manual
  • InterfaceRS-232
  • WaterproofYes
  • Battery4 x AA batteries (included)
  • Width (cm)8.255
  • Height (cm)18.415
  • Width (in)3 1/4
  • Length (in)2 1/4
  • Height (in)7 1/4
  • DescriptionWaterproof Handheld Meter Only
  • Warranty3 year
3 Year WarrantyConformite EuropeenneIP 67ISO9001:2000 Certified Supplier

Key Features

  • Measure pH or ISE with 0.001 maximum resolution
  • Out-of-date or unperformed calibrations are now things of the past!
  • Don't waste time looking for those cables!
  • Use in dirty and wet environments
  • 3 year warranty

More About this Item

These waterproof handheld meters feature a –2.00 to 20.00 pH range with 0.001 pH resolution. Combined with accuracies up to ±0.002 pH, these meters provide a new benchmark in our pH meter line. Store up to 500 data sets in the meter‘s memory. Time-and-date stamping meets Good Laboratory Practice (GLP) standards.

The pH 620 meters offer the latest in pH calibration capabilities including up to six calibration points, a user-settable “calibration due” alarm, and up to 15 buffer options with auto-buffer recognition of USA, NIST, DIN, and PWB standards.

The infrared (IrDA) wireless technology makes PC downloading more convenient and easier. Dual powerup allows you to use batteries while out in the field or the optional AC power adapter while in the lab.

As always, our waterproof meters are IP67 rated for protection against both dust and water, even with no probes attached.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat