This item is now discontinued. We recommend the following as suggested replacements: WZ-35660-78

Oakton PD 650 Waterproof Handheld Meter Kit

Representative image only.
Oakton Item # WZ-35432-70
Top-of-the-line dual parameter meters offer features typically found on much more expensive models!
  • Simultaneously measure your choice of pH, mV, or ISE and dissolved oxygen
  • Log up to 500 data points with GLP compliance
  • Wireless IrDA communication for truly waterproof interfacing capabilities
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 6 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min ISE0 to 19999 ppm
  • ISE Resolution0.001 ppm; 0.001 molar
  • ISE Accuracy±0.5% of reading
  • Min Dissolved Oxygen0 to 50 ppm, 0 to 600% saturation
  • Dissolved Oxygen Accuracy±2%, or ±0.2 ppm
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Min Temperature (° F)14
  • Max Temperature (° F)230
  • Temperature compensationAutomatic or manual
  • InterfaceIrDA and RS-232C
  • Data Logging (points)500 data sets with time-date stamping
  • WaterproofYes
  • Battery4 x AAA batteries (included)
  • Width (cm)8.255
  • Height (cm)18.415
  • Width (in)3 1/4
  • Length (in)2 1/4
  • Height (in)7 1/4
  • DescriptionWaterproof Handheld Meter Kit
  • Warranty3 year
3 Year WarrantyConformite EuropeenneIP 67ISO9001:2000 Certified Supplier

Key Features

  • Simultaneously measure your choice of pH, mV, or ISE and dissolved oxygen
  • Log up to 500 data points with GLP compliance
  • Wireless IrDA communication for truly waterproof interfacing capabilities
  • 3 year warranty

More About this Item

These 650 meters feature user-selectable “calibration due” alarms, automatic or manual standards recognition, and single or multiple calibration points — providing flexibility in meeting your quality objectives. The large backlit LCD shows the measured value, measured units, temperature, time, battery life, and menu options. Infrared (IrDA) wireless technology lets you easily download data without any cables.

PD 650 meters measure pH and ISE with a 0.001 maximum resolution. The PD 650 measures oxygen saturation and concentration using a galvanic dissolved oxygen probe.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat