Salinity Resolution0.01 or 0.1 ppm; 0.001 or 0.01 ppt
Salinity Accuracy±1.0% FS
Min Resistivity2.0 Ohm
Max Resistivity20.0 MOhm
Resistivity Resolution0.001, 0.01, or 0.1 Ω; 0.1 kΩ; 0.001, 0.01, or 0.1 MΩ
Resistivity Accuracy±1% FS
Min Dissolved Oxygen0 to 50 ppm, 0 to 600% saturation
Dissolved Oxygen Accuracy±2%, or ±0.2 ppm
Min Temperature (° C)-10
Max Temperature (° C)45
Min Temperature (° F)14
Max Temperature (° F)113
Temperature compensationAutomatic or manual
InterfaceIrDA and RS-232C
Data Logging (points)500 data sets with GLP
WaterproofYes
Battery4 x AA batteries (included)
Width (cm)8.255
Height (cm)18.415
Width (in)3 1/4
Length (in)2 1/4
Height (in)7 1/4
Accessories IncludedMeter, probes, carrying case, calibration pouches, and NIST-Traceable calibration
DescriptionWaterproof Multiparameter Meter Kit with NIST Cal Cert
Warranty3 year
Key Features
No more excuses for missing data! Out-of-date or unperformed calibrations are now things of the past!
Don’t waste time looking for those cables!
Use in dirty and wet environments
3 year warranty
More About this Item
This top-of the-line waterproof meter offers features that are typically found on much more expensive models. The IP67-rated housing allows use in dirty and wet environments without harm to the meter. Datalog up to 500 points with time-and-date stamp. User-selectable “calibration due” alarms, automatic or manual standards recognition, and single or multiple calibration points provide flexibility in meeting your data quality objectives. Infrared (IrDA) wireless technology lets you easily download data — no need for additional cords. The large backlit LCD shows the measured value, measured units, temperature, time, battery life, and menu options.
The pH range is from –2.000 to 20.000 with accuracies up to ±0.002. The pH calibration capabilities include up to six calibration points, a user-settable “calibration due” alarm, and up to 15 buffer options with auto-buffer recognition of USA, NIST, DIN, and PWB standards.
Meter measures conductivity (up to 500 mS), TDS, salinity, or resistivity with either a 2-or 4-electrode conductivity cell. Linear or pure-water temperature compensation curve provides accurate data at all levels of conductivity. Also features set point alarms to indicate when concentrations are outside of programmed limits.
Meter determines oxygen saturation up to 600% and concentration up to 90 mg/L. The galvanic dissolved oxygen probe does not require any warm-up time so you can take readings immediately. Salinity correction is automatically compensated for after manual input of concentration. Barometric pressure is automatically compensated for by the included barometer.
Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.