Oakton Ion Selective Electrode (ISE), Double-Juntion, Glass; Water Hardness
Representative image only.
Oakton Item # WZ-35802-49
A wide variety of ion selective electrodes are availabe to suit a wide array of applications
  • Epoxy-body types are either single- or double-junction
  • Glass-body type is double junction
  • 1 year warranty
PRODUCT OPTIONS
£556.72GBP / Each
£668.06GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction, membrane
  • Electrode Body MaterialGlass
  • Ion-Selective ElectrodeWater hardness Ca/Mg
  • ConnectorBNC
  • DescriptionIon Selective Electrode (ISE), Double-Juntion, Glass; Water Hardness
  • Warranty1 year
1 Year Warranty
Key Features
  • Epoxy-body types are either single- or double-junction
  • Glass-body type is double junction
  • 1 year warranty
More About this Item
Select from four electrode types: membrane, solid-state, gas sensing, and glass bulb; typical response time is 20 to 30 seconds.