Oakton Hygrothermograph Chart Recorder Paper; 22 to 104°F, 7 Day, 2 Hour; 100/Pack
Representative image only.
Oakton Item # WZ-08369-61
Replacement paper for Oakton® hygrothermograph chart recorders
  • Varying temperature ranges and speeds
PRODUCT OPTIONS
£49.30GBP / Pkg of 100
£59.16GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeDrum
  • Chart Speed7 day
  • Chart Width (in)3.625
  • Chart Length (in)8.1875
  • Max Temperature (° F)104
  • Min Temperature (° F)22
  • DescriptionHygrothermograph Chart Recorder Paper; 22 to 104°F, 7 Day, 2 Hour; 100/Pack
Key Features
  • Varying temperature ranges and speeds
More About this Item
Choose the paper compatible with your chart recorder and application needs.