Oakton High-Accuracy Buffer Solution, pH 10.000; 500 mL

Representative image only.
Oakton Item # WZ-05942-69
pH Calibration buffers for tight tolerances
  • For meters with 0.001 resolution
  • Precision reference standard: pH 10.000 ±0.005 at 25°C
  • 18 month warranty

PRODUCT OPTIONS

£38.36GBP / Each
£46.03GBP (incl VAT)
Choose the buffer value and volume that fits your needs
Oakton® pH Buffer Solutions
£15.83 - £166.40GBP / Pint of 1
£19.00 - £199.68GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Value10
  • Buffer Kit Values10.000 ±0.005 at 25°C
  • Volume (mL)500
  • ColorBlue
  • DescriptionHigh-Accuracy Buffer Solution, pH 10.000; 500 mL
  • Warranty18 months
18 Month Warranty

Key Features

  • For meters with 0.001 resolution
  • Precision reference standard: pH 10.000 ±0.005 at 25°C
  • 18 month warranty

More About this Item

Oakton® High-accuracy pH buffer solutions are sold in 500 mL glass bottles, individually or as a case of twelve.
Buffers are color-coded for easy visual identification.

Store away from direct sunlight.

Certificate of Analysis

Enter lot number
or click to view a Sample C of A

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat