This item is now discontinued. We recommend the following as suggested replacements: WZ-56700-15

Oakton ECPHCP0550 pH 550 pH/orp Monitor

Representative image only.
Oakton Item # WZ-56717-30
Small size, economical monitor with display
  • Measure and display both pH/ORP and temperature
  • Automatic or manual temperature compensation
  • Asymetrical or symmetrical mode operation
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Control TypeOn/Off
  • Display TypeLCD
  • Housing MaterialIP65
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01
  • pH Accuracy±0.01 pH unit
  • Min mV (mV)-1000
  • Max mV (mV)1000
  • mV Resolution1mV
  • mV Accuracy±2mV
  • Min Temperature (° C)-10
  • Max Temperature (° C)125
  • Min Temperature (° F)14
  • Max Temperature (° F)257
  • Temperature Resolution0.1°
  • Temperature Accuracy±0.5°C
  • Temperature CompensationAutomatic or Manual
  • Width (cm)9.53
  • Height (cm)9.53
  • Width (in)3 3/4
  • Height (in)3 3/4
  • Depth (in)2 53/64
  • DescriptionpH 550 pH/orp Monitor
  • Warranty1 year
1 Year WarrantyConformite Europeenne

Key Features

  • Measure and display both pH/ORP and temperature
  • Automatic or manual temperature compensation
  • Asymetrical or symmetrical mode operation
  • 1 year warranty

More About this Item

The pH 550 monitors are used to measure pH /ORP and temperature values in applications such as water treatment and monitoring, chemical processing, or food processing. The monitors have user-friendly features like menu-driven programs and calibrations that are easily performed with the push-button keypad. All setup and calibration parameters are retained in the nonvolatile memory to prevent data loss during power outages.

Measurements are compensated for temperature variations automatically when used with pH electrodes that have temperature elements. The monitors also offer manual temperature compensation with independent settings for calibration and process temperature. One- or two-point calibrations can be performed with USA or NIST buffer sets. All values, operational, and error messages are indicated on the large, dual-line LCD. All setup and calibration parameters are retained in the nonvolatile memory to prevent data loss during power outages.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat