Environmental Express EC200 Conductivity Cell, K=1; 1-m Cable
Representative image only.
Environmental Express Item # WZ-35660-58
Enhance the versatility of your Oakton 200 series meters
  • Suitable for use with Oakton 200 series meters and meter kits
  • 6 month warranty
PRODUCT OPTIONS
£276.64GBP / Each
£331.97GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • ConnectionsBNC and phono
  • Min Conductivity0.1 mS/m
  • Max Conductivity10 S/m
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • Min Temperature (° F)32
  • Max Temperature (° F)176
  • Length (cm)15
  • Diameter (cm)1.6
  • Length (in)5 29/32
  • DescriptionConductivity Cell, K=1; 1-m Cable
  • Warranty6 month
6 Month WarrantyConformite Europeenne
Key Features
  • Suitable for use with Oakton 200 series meters and meter kits
  • 6 month warranty
More About this Item
For your convenience, cables, electrodes, and replacement parts are available for your Oakton meters.