Environmental Express EC 100 Portable Conductivity Meter with EC/ATC Probe

Representative image only.
Environmental Express Item # WZ-35604-10
Convenient, compact design with large display
  • Measured parameter and temperature shown on display at same time
  • Stability, calibration, and low-battery indicators
  • Waterproof and dust-resistant IP57 rating – great for use under harsh environments
  • 2 year warranty

PRODUCT OPTIONS

£451.75GBP / Each
£542.10GBP (incl VAT)

£187.00 - £208.21GBP / Each
£224.40 - £249.85GBP (incl VAT)

for an additional
£187.00GBP / Each
£224.40GBP (incl VAT)
for an additional
£208.21GBP / Each
£249.85GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeConductivity, TDS, salinity, temperature
  • Display typeLCD
  • ConnectionsBNC & Φ2.5 socket
  • Buffer RecognitionAutomatic standard and user-defined conductivity calibration solutions
  • Calibration1-4 points automatic
  • Min Conductivity0 mS/cm
  • Max Conductivity0.00 to 19.99 μS/cm, 20.0 to 199.9 μS/cm,200 to 1999 μS/cm; 2.00 to 19.99 mS/cm, 20.0 to 199.9 mS/cm
  • Conductivity Resolution0.01, 0.1, 1 μS/cm; 0.01, 0.1 mS/cm
  • Conductivity Accuracy±1.0% full scale
  • CellK=1.0
  • Cell Constant0.1, 1, 10 cm–1
  • Min TDS0 g/L
  • Max TDS100 μg/L
  • TDS Accuracy±1.0% full scale
  • Min Salinity0 ppt
  • Max Salinity100 ppt
  • Salinity Accuracy±1.0% full scale
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Temperature Resolution0.1°F/°C
  • Temperature Accuracy±0.9°F (0.5°C) ±1 digit
  • Temperature compensation0 to 100˚C (manual or automatic)
  • Temperature Coefficient2.0%/°C
  • WaterproofIP57
  • Battery3 AA (included)
  • Battery life (hr)400
  • Length (cm)9.1
  • Width (cm)19
  • Height (cm)3.3
  • Width (in)7 31/64
  • Length (in)3 37/64
  • Height (in)1 19/64
  • Stirring ControlNo
  • NIST Calibration IncludedNo
  • Probe IncludedEC/temperature (ATC)
  • Accessories IncludedNone
  • DescriptionOakton EC 100 Portable Conductivity Meter with EC and ATC probes
  • Warranty2 year
Two year warrantyConformite Europeenne

Includes

EC 100 portable conductivity meter, plastic conductivity electrode, and temperature probe.

Key Features

  • Measured parameter and temperature shown on display at same time
  • Stability, calibration, and low-battery indicators
  • Waterproof and dust-resistant IP57 rating – great for use under harsh environments
  • 2 year warranty

More About this Item

The new addition to the Environmental Express Oakton product line features updates to measuring water quality easier and faster. The larger display is 2.75 in. by 2.0 in. for better viewing angle, and has indicators for battery life, stability, and calibration. It also shows both the parameter and temperature measurements at the same time. The EC 100 meter measures conductivity, TDS, salinity, and temperature.

The built-in microprocessor chip enables advanced functions, such as auto calibration, auto temperature compensation, parameter setting, self-diagnosis, auto power-off, and low-battery reminders. The meter uses advanced digital processing technology, intelligently improving the response time and accuracy of your measurements. Stable reading and auto lock display modes are available.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat