Environmental Express DO 100 Replacement Electrode
Representative image only.
Environmental Express Item # WZ-35643-01
Quality accessories for Oakton DO 100 and 1000 meters
  • Choose from a variety of accessories for Oakton meters
  • Compatible with DO 100 meters (35643-00 & 35643-05)
  • Integrated temperature and salinity sensors
  • Regular electrode polarization only takes 5 minutes
  • Quick response –– measurement reaches 90% response within 30 seconds
  • 6 month warranty
PRODUCT OPTIONS
£311.97GBP / Each
£374.36GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeElectrode
  • MaterialABS plastic body, PTFE membrane
  • DescriptionDO 100 Replacement Electrode
  • Warranty6 month
6 Month Warranty
Key Features
  • Choose from a variety of accessories for Oakton meters
  • Compatible with DO 100 meters (35643-00 & 35643-05)
  • Integrated temperature and salinity sensors
  • Regular electrode polarization only takes 5 minutes
  • Quick response –– measurement reaches 90% response within 30 seconds
  • 6 month warranty
More About this Item
For your convenience, electrodes, membrane caps, and other accessories are available for your Oakton meters.
Temperature sensor range is from 32 to 113°F (0 to 45°C). The electrode has a 3 ft cable.