Oakton Conductivity Calibration Pouches, 84 uS; 20/Pk
Representative image only.
Oakton Item # WZ-35653-08
Sealed for freshness and easy travel
  • Tear off a corner, or the entire top—opens wide enough for all probe types
  • Notched corners for easy opening from either side
  • Calibrate to 84 µS with these one-use pouches
  • 6 month warranty
PRODUCT OPTIONS
£52.46GBP / Pkg of 20
£62.95GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Conductivity Value (µS)84
  • Volume (mL)20
  • Volume (oz)0.7
  • Volume20 pouches
  • Conductivity-to-TDS Calibration Values (ppm 442)50.5
  • Conductivity-to-TDS Calibration Values (ppm KCl)40.38
  • Conductivity-to-TDS Calibration Values (ppm NaCl)38.04
  • DescriptionConductivity Calibration Pouches, 84 uS; Box of 20 Pouches
  • Warranty6 month warranty
6 Month Warranty
Key Features
  • Tear off a corner, or the entire top—opens wide enough for all probe types
  • Notched corners for easy opening from either side
  • Calibrate to 84 µS with these one-use pouches
  • 6 month warranty
More About this Item
Single-use 20-mL pouches are sealed to ensure freshness—take these convenient standard solutions anywhere! Don't worry about transferring solution into another container—calibrate directly from the pouch.
A µS-to-ppm conversion table is printed on each pack for quick reference.