NSI Lab Solutions NSIQCI-055 Universal Wastewater Solid Standard; 4 x 1 L bottles
Representative image only.
NSI Lab Solutions Mfr # NSIQCI-055 – Item # WZ-35200-27
Certified for TDS, TS, and TSS test methods
  • Ready to use without dilution — simply shake and pour
  • Store at room temperature — no need to refrigerate
  • Certificate of Analysis provides true values and acceptance limits
  • Comes as a case of 4 x 1-liter bottles
£129.07GBP / Pkg of 4
£154.88GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • ClassStandards
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)4000
  • Weight (mL)4000
  • Packaging4000 mL
  • DescriptionUniversal Wastewater Solid Standard; 4 x 1 L bottles
Key Features
  • Ready to use without dilution — simply shake and pour
  • Store at room temperature — no need to refrigerate
  • Certificate of Analysis provides true values and acceptance limits
  • Comes as a case of 4 x 1-liter bottles