This item is now discontinued. Please browse similar items
NPS Spilfyter Mercury Amalgamation Powder, 500 g Shaker Bottle
Representative image only.
NPS Mfr # 520500 – Item # WZ-86424-58
Convert mercury spill to an amalgam, preventing mercury vapors
SPECS & DESCRIPTION
Specifications & Description
  • Description500 g shaker bottle
More About this Item
Amalgamation powder converts mercury into an amalgam, or paste-like consistency. This prevents mercury vapors from being emitted. Absorbs up to 15 g of mercury/100 g of powder.