NPS Spilfyter Kolor-Safe Acid Neutralizer, 1 gallon
Representative image only.
NPS Mfr # 410004 – Item # WZ-86423-11
Three-color change lets you know when spill is completely neutralized
PRODUCT OPTIONS
£60.78GBP / Gallon of 1
£72.94GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAcid Neutralizer, 1 gallon
More About this Item
Protect your workers and facility from acid spills by quickly and efficiently neutralizing them. Simply apply the neutralizer until the spill goes through the three-color change. Acid neutralizer changes from purple to yellow to a reddish color to indicate total neutralization.