NPS Spilfyter Compact Formaldehyde Spill Kit
Representative image only.
NPS Mfr # 481001 – Item # WZ-86424-56
Comply with OSHA regulations by controlling and cleaning formaldehyde spills
  • Kit includes everything you need for small formaldehyde spills
£131.31GBP / Each
£157.57GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCompact Formaldehyde Spill Kit
Key Features
  • Kit includes everything you need for small formaldehyde spills
More About this Item
The formaldehyde solidifier suppresses the vapor as well as solidifies the liquid for easy disposal.
Note: The resulting solid is still hazardous and must be disposed of in adherence to all local, state, and federal regulations.