NPS Spilfyter Chemical Spill Classifier Kit; 6 Tests/Kit
Representative image only.
NPS Mfr # 570001 – Item # WZ-86423-00
Classify unknown spills in 30 seconds
PRODUCT OPTIONS
£93.64GBP / Each
£112.37GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionChemical Spill Classifier Kit; 6 Tests/Kit
Includes
Kit includes six test strips, one bobbin of thread, one color reference chart, and one plastic tube with cap. Additional test strips and charts may also be ordered separately.
More About this Item
Convenient strips change color to identify risk type and classify spill for proper treatment. Compare color indicator to laminated, reusable reference chart to classify unknown liquid.
Chemical classifier kit tests for acid or base, iodine, oxidizers, chlorine, fluoride, bromine and petroleum/organic products.