This item is now discontinued. Please browse similar items
NOSHOK 850-1-8-300/1100-2-090-6 Temperature Switch, -300/1100, 9" .Stem Electronic Temperature Switch
Representative image only.
NOSHOK Mfr # 850-1-8--4/176-2-250mm-6 – Item # WZ-39007-60
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTemperature Switch, -300/1100, 9\" .Stem Electronic Temperature Switch
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
The NoShok 850 temperature switch measures and displays temperature and has one or two switching outputs as well as an optional analog output. The temperature set points, reset points, switching functions and the measuring range of the optional analog output are simple to adjust via two buttons. All these features and measuring range between -300F and 1100F (-200C and 600C) cover the majority the majority of temperature measuring and switching tasks.
  • Compact dimensions
  • Simple handling
  • Cost effective
  • Service-friendly
  • Customized solutions