This item is now discontinued. Please browse similar items
Norwell Dipstik Reusable, FEP bailer, 1-7/8"OD X1', with standard top/bottom
Representative image only.
Norwell Dipstik Mfr # D8653000 – Item # WZ-05497-00
Sink fast for quicker sampling
  • Available in two styles a nd lengths
  • Order polyester suspension cord separately
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialFEP, PTFE
  • Cap ConfigurationStandard/ standard
  • Length (in)12
  • Length (ft)1
  • Length (cm)30.48
  • DescriptionReusable, FEP bailer, 1-7/8\"OD X1', with standard top/bottom
  • Warranty1 year
1 Year Warranty
Key Features
  • Available in two styles a nd lengths
  • Order polyester suspension cord separately
  • 1 year warranty
More About this Item
FEP Reusable Bailers are available in two styles and lengths. Economy bailers have a 1-3/4" OD and feature a fixed top and bottom ball valve. Standard bailers have a 1-7/8" OD and feature interchangeable top and bottom cap configurations. Standard bailers come in six of the most popular top/bottom cap configurations.