This item is now discontinued. Please browse similar items
Norwell Dipstik D8653276 polypropylene drum and tank sampler, 3.5'long
Representative image only.
Norwell Dipstik Mfr # D8653276-1 – Item # WZ-99200-10
No leachable fillers or plasticizers
  • No glass parts to break for added safety and protection Easily identify layers through translucent material Easy to clean
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Descriptionpolypropylene drum and tank sampler, 3.5'long
  • Warranty6 month
6 Month Warranty
Key Features
  • No glass parts to break for added safety and protection Easily identify layers through translucent material Easy to clean
  • 6 month warranty
More About this Item
Sample free-flowing liquids from almost any source. Safety pour top permits safe and neat transfer of contents. Push/seal bottom snaps closed easily; opens by hand or with the safety opening tool sold separately. Polypropylene (PP) samplers have heat-welded bottom caps.