North by Honeywell 8B1532/9Q Butyl drybox gloves, 15 mil, size 9-3/4, 1 pair
Representative image only.
Honeywell Mfr # 8B1532/9Q – Item # WZ-86569-20
Shoulder-length, seamless gloves for critical working conditions
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • MaterialButyl
  • Thickness (mil)15
  • SizeMedium
  • Length (in)32
  • Length (cm)81
  • ColorBlack
  • SterileNo
  • DescriptionButyl drybox gloves, 15 mil, size 9-3/4, 1 pair
More About this Item
Butyl gloves allow highest impermeability to water, vapor and geses. They maintain flexibility even at low temperatures and are resistant to oxygenated solvents and most oxidizing chemicals.