This item is now discontinued. Please browse similar items
North by Honeywell 125LNR Nitrile Finger Cots, Medium, 720/bag
Representative image only.
North by Honeywell Mfr # 125LNR – Item # WZ-81605-92
Free from latex proteins that may cause allergic reactions
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)3
  • SizeMedium
  • Length (in)2 1/4
  • Length (cm)6
  • ColorWhite
  • SterileNo
  • DescriptionNitrile Finger Cots, Medium, 720/bag
More About this Item
Powder-free 100% nitrile formulation offers superior stretch and comfort to reduce finger fatigue and encourage user acceptance. These 3-mil thick cots have inherent anti-static properties — eliminating damaging static discharge in many applications. Rolled edges ensure quick and easy donning.