This item is now discontinued. Please browse similar items
Newtex Industries Style 130 Gloves with Leather Palm, 1 pr
Representative image only.
Newtex Industries Mfr # 2100009 – Item # WZ-86364-32
Reinforced palms provide excellent abrasion and cut protection
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialLeather
  • SizeOne size fits all
  • ColorWhite
  • ComplianceEN 407
  • ApprovalUL-listed
  • SterileNo
  • DescriptionGloves with Leather Palm, 1 pr
  • Warranty1 month
1 Month Warranty
Key Features
  • 1 month warranty
More About this Item
Reinforced gloves offer heat protection up to 800°F (427°C). Reinforced palms are sewn-in and double-stitched for maximum durability. Select aramid palm gloves for additional cut protection. Leather palm gloves feature superior abrasion resistance. One size fits all. Made in the USA.
Approval: UL listed
Compliance: European EN 407 industrial standard