This item is now discontinued. Please browse similar items
Nederman High-Efficiency Fume Extractor Arm with Light, 120"L
Representative image only.
Nederman Mfr # 1055-4335,-1235,14513637 – Item # WZ-33700-12
Up to 650 cfm air capacity for particles, smoke, and dust
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Switches2
  • Power (VAC)115 / 230
  • Power (Hz)50/60
  • Exhaust Volume (CFM)650
  • DescriptionHigh-Efficiency Fume Extractor Arm with Light, 120\" L
  • Warranty3 Years
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Fume extractor arms are perfect for welding booths and other industrial applications requiring large air capacities. Arms come in four lengths: 7 ft; 10 ft; 14 ft; and telescopic, which measures 34" when retracted, 5 ft when fully extended. Telescopic and 7-ft arms are suitable for small work areas; 10-ft and 14-ft arms are best for wider work areas. Specially designed friction joints allow hoods to be positioned at any point within the work area. Arms are supported by a 360deg swivel elbow for unlimited maneuverability. Maximum operating temperature is 158degF (70degC). Assembly is required for all models.
The 7-ft, 10-ft, and 14-ft arms include 10" x 12" oval polycarbonate hood with handle and damper, and 6-1/4" OD PVC fiberglass-reinforced hose. Telescopic a