This item is now discontinued. Please browse similar items
Nasco Sampling B01096WA Extendable Tank Sampler, 10-ft total length
Representative image only.
Nasco Sampling Mfr # B01096WA – Item # WZ-99155-20
Sample at any depth with 5-ft section additions
  • Constructed of 3/4" diameter durable butyrate plastic pipe
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionExtendable Tank Sampler, 10-ft total length
  • Warranty1 year
1 Year Warranty
Key Features
  • Constructed of 3/4" diameter durable butyrate plastic pipe
  • 1 year warranty
More About this Item
Sampler includes two, 5-foot sections and holds approximately three ounces (80 mL) per foot. Add additional sections as needed.