This item is now discontinued. Please browse similar items
Motic Instruments Moticam X Digital Microscope Camera, Wi-Fi, 1/3" CMOS
Representative image only.
Motic Instruments Mfr # 1100600100691 – Item # WZ-03906-18
Fast plug-and-play cameras relay images from your microscope to your computer or monitor
  • Supplied software measures specimens and logs each image with automatic reference number, date, and time
SPECS & DESCRIPTION
Specifications & Description
  • Resolution2.0 MP
  • Eyepiece Diameter (mm)12
  • Image Sensor1/3\" CMOS
  • OutputWi-Fi
  • System RequirementsWindows® XP and higher, Mac OSX®
  • DescriptionDigital Microscope Camera, Wi-Fi, 1/3\" CMOS
ISO9001 Certified Supplier
Key Features
  • Supplied software measures specimens and logs each image with automatic reference number, date, and time
More About this Item
Capture electronic images and videos by converting your compound or stereo microscope into a powerful digital imaging system. These affordable digital cameras capture still or video images and instantly transmit them for real-time full screen viewing. Timed-interval shots provide time lapse observation—save, manipulate, print, and send!
Rapid view algorithms let you display at a faster frame rate while still being able to capture images at the highest resolution. These cameras are also great for capturing images of objects too large for a microscope stage. The included macro viewing tube is a stand that sits on the bench and holds the camera above a specimen.
Moticam X (Wi-Fi) sends streaming images to up to six Wi-Fi enabled devices such as tablets, smart phones, or computers. Download the free app to use iOS or Android; load the inlcuded software for use with OSX or Windows®-based computers. Also includes a power supply.