This item is now discontinued. Please browse similar items
Motic Instruments Moticam 5 Digital Microscope Camera, 1/2.5" CMOS
Representative image only.
Motic Instruments Mfr # 1100600100631 – Item # WZ-03906-15
Fast plug-and-play cameras relay images from your microscope to your computer or monitor
  • Supplied software measures specimens and logs each image with automatic reference number, date, and time
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Resolution5 MP
  • Eyepiece Diameter (mm)12
  • Image Sensor1/2.5\" CMOS
  • OutputUSB
  • System RequirementsWindows® XP and higher, Mac OSX®
  • Cable(s)USB
  • Power5 VDC via USB
  • DescriptionDigital Microscope Camera, 1/2.5\" CMOS
  • Warranty1 Year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Supplied software measures specimens and logs each image with automatic reference number, date, and time
  • 1 year warranty
More About this Item
Capture electronic images and videos by converting your compound or stereo microscope into a powerful digital imaging system. These affordable digital cameras capture still or video images and instantly transmit them for real-time full screen viewing. Timed-interval shots provide time lapse observation—save, manipulate, print, and send!
Rapid view algorithms let you display at a faster frame rate while still being able to capture images at the highest resolution. These cameras are also great for capturing images of objects too large for a microscope stage. The included macro viewing tube is a stand that sits on the bench and holds the camera above a specimen.
Moticam 5 features the highest resolution of 10 megapixels, giving you the ability to convert images into 300 dpi files—ideal for documentation purposes!