This item is now discontinued. Please browse similar items
Motic Instruments 282A Digital Microscope Camera, 2/3" CCD
Representative image only.
Motic Instruments Mfr # 282A – Item # WZ-03908-73
Capture high-quality images with this scientific-grade CCD camera
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Resolution2580 x 1944
  • Image Sensor2/3\" CCD
  • System Requirements1 GB RAM or better, Windows® XP or better, Mac® OSX
  • PowerUSB
  • DescriptionDigital Microscope Camera, 2/3\" CCD
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Internal 4-frame buffer provides faster and more consistent frame rates. On-board image processing allows you to apply white balance or color corrections before the image is sent to your computer. Capture live, still, time-lapse, and video images with the included Motic Images Advanced 3.2 software. Software features include measurement, editing, amalgamation, manual/single point segmentation, single image assembly from multiple images, and multi-language display. USB 2.0 output provides easy communication with your PC or laptop and quick plug-and-play setup.