This item is now discontinued. We recommend the following as suggested replacements: WZ-04775-65
Cole-Parmer Essentials Standard Stir Bar with Spinning Ring, 2"L x 3/8" dia
Representative image only.
Cole-Parmer Essentials Item # WZ-08552-00
Resist spin out at high speeds
  • Super-strong alnico V magnets remain coupled to drive magnets
  • One-piece coating of PTFE resists corrosion, heat, and cold
  • Chemically inert stir bars won't contaminate liquid being stirred
  • Guaranteed to be leakproof
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)2
  • Length (cm)5
  • Diameter (in)0.375
  • Diameter (cm)0.9525
  • DescriptionStandard Stir Bar with Spinning Ring, 2\"L x 3/8\" dia
  • Warranty2 year
Two year warranty
Key Features
  • Super-strong alnico V magnets remain coupled to drive magnets
  • One-piece coating of PTFE resists corrosion, heat, and cold
  • Chemically inert stir bars won't contaminate liquid being stirred
  • Guaranteed to be leakproof
  • 2 year warranty
More About this Item
These pivot ring PTFE stir bars reduce the contact area between stir bar and vessel, reducing friction and noise. Low-friction PTFE coating ensures smooth spinning and maximum energy transfer.