This item is now discontinued. Please browse similar items
Monarch DC2C1-U4-10000 4CH.Rcdr Active Color Display Recorder Paperless
Representative image only.
Monarch Mfr # DC2C1-U4-10000 – Item # WZ-49005-77
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels4
  • Chart TypeDigital
  • Min Temperature (° F)-150
  • Max Temperature (° C)4350
  • Min Temperature (° C)-101
  • Power (VAC)100 to 240
  • Description4CH.Rcdr Active Color Display Recorder Paperless
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
FEATURES
  • 2, 4, 6 or 12 direct universal inputs: DC voltage, current, thermocouples, RTD and dry contact
  • Serial output RS232 or RS485 (optional)
  • Ethernet 10 Base T option available
  • High speed storage capability: up to 8 samples per second/channel
  • Storage Media Compact Flash Card
  • 5.6" Brilliant TFT active matrix color display or 5" monochrome display
  • Intuitive Touchscreen Control with anti-glare coating
  • 6 relay contact or solid state relay outputs/3 digital control input
  • Parallel printer output (optional)