LaMotte 3002 Test Strips, Chlorine Dioxide, 0-500ppm, 50 Strips/Vial 99532-34
Representative image only.
LaMotte Mfr # 3002 – Item # WZ-99532-34
Use these chemically treated paper strips as a quick test for sanitizer strength
  • Compare color change to included chart for easy-to-read results
PRODUCT OPTIONS
£14.24GBP / Each
£17.09GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • TestChlorine dioxide
  • Reagent PlatformStrips
  • Tests Per Kit50
  • DescriptionTest Strips, Chlorine Dioxide, 0-500ppm, 50 Strips/Vial 99532-34
Key Features
  • Compare color change to included chart for easy-to-read results
More About this Item
Strips and color chart come supplied in a waterproof plastic vial.
Test strips permit an easy swirl and read way to determine chlorine dioxide levels from 0 to 500ppm.
Call Us At+44 (0)1480 272279
Live chat
Alternatives
Alternatives