Minntech 279-30-011 Hollow Fiber Filter Capsule, 0.5-2.5L, 0.2um, 1/4"NPTM; 6/Pk
Representative image only.
Minntech Mfr # 279-30-011 – Item # WZ-29880-22
Provide high flow rates with low pressure drop
  • Steam sterilize capsules in-line or autoclave off-line
  • Polysulfone membrane offers low protein binding
  • Capsules have a polycarbonate case and end caps and polyurethane end seals
  • 1 year warranty
PRODUCT OPTIONS
£1,151.73GBP / Case of 6
£1,382.08GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPolysulfone
  • Pore Size (micron)0.2
  • Housing MaterialPolycarbonate
  • Connections1/4\" NPT(M)
  • Effective Filtration Area (cm²)1858
  • Surface Area (m²)0.1858
  • Typical Water Flow Rate0.5 to 2.5 LPM
  • Max Pressure (PSI)30
  • Max Pressure2
  • DescriptionHollow Fiber Filter Capsule, 0.5-2.5L, 0.2um, 1/4\"NPTM; 6/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Steam sterilize capsules in-line or autoclave off-line
  • Polysulfone membrane offers low protein binding
  • Capsules have a polycarbonate case and end caps and polyurethane end seals
  • 1 year warranty
More About this Item
These new hollow fiber filters offer superior performance in a compact size. Filters have up to six times more surface area compared to other pleated membrane filters, providing greater capacity and longer filter life. The unique membrane structure helps remove bacteria, high-level endotoxins, pyrogens, and spores — ideal for a variety of water purification applications. Maximum differential pressure is 30 psi (2 bar). Capsules and cartridges provided nonsterile.